Learn More
Medchemexpress LLC 3-Indolyl acetate | 608-08-2 | 99.3% | 175.18 | 10 G
SDP
Supplier:  Medchemexpress LLC HY13788410G
Encompass_Preferred
3-Indolyl acetate is an ester derivative of indole that is ubiquitous in various plant tissues, especially in certain cruciferous vegetables such as broccoli and cabbage. It acts as an important signaling molecule in plants, regulating physiological processes such as growth and defense against pathogens. It also has potential applications in the pharmaceutical industry, as it has been shown to have antitumor and anti-inflammatory effects in some studies. As a biochemical reagent, it can be used as a biological material or organic compound for life science related research.
  • Ester derivative of indole
  • Found in various plant tissues
  • Regulates plant growth and defense mechanisms
  • Shows potential antitumor effects
  • Shows potential anti-inflammatory effects
  • Suitable for life science-related research
Catalog No. 50-003-50684