Learn More
Medchemexpress LLC 3-iodo-L-tyrosine | 70-78-0 | 307.09 | 5 G

Supplier: Medchemexpress LLC HYW0084525G
H-Tyr(3-I)-OH is a potent and effective inhibitor of tyrosine hydroxylase. This compound serves as an intermediate in the biosynthesis of thyroid hormones and functions as a metabolite in both human and mouse biological systems.
- Potent and effective tyrosine hydroxylase inhibitor.
- Acts as an intermediate in thyroid hormone production.
- Functions as a human and mouse metabolite.
- High purity of 99.93%.
- Powder form is stable for 3 years when stored at -20°C.
- Solution form is stable for 6 months at -80°C or 1 month at -20°C.
- Shipped at room temperature within the continental US.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.