Learn More
Sigma Aldrich Fine Chemicals Biosciences 3 Isobutyl 1 methylxanthin5G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences I58795X5G
Encompass
3-isobutyl-1-methylxanthine/IBMX is a non-selective non-specific inhibitor of cAMP and cGMP phosphodiesterases. IBMX can induce melanogenesis and can be used as a positive control in melanogenesis research. In oocyte research IBMX assists in maintaining the germinal vesicle (GV) arrest of prophase I oocytes.
Catalog No. 50-290-8084