Learn More
Medchemexpress LLC 3-Isoquinolinecarboxylic acid | 6624-49-3 | 173.17 | 10 G
SDP
Supplier:  Medchemexpress LLC HY4100410G
Encompass_Preferred
3-Isoquinolinecarboxylic acid is a synthetic intermediate that has been used in the design, synthesis, and testing of novel anti-tumor leads. It is suitable for use as a laboratory chemical and in the manufacture of substances.
  • Used to design antitumor compounds.
  • Synthetic intermediate.
  • Suitable for laboratory chemical applications.
  • Suitable for the manufacture of substances.
Catalog No. 50-004-04677