Learn More
Medchemexpress LLC 3-Maleimidopropionic acid | 7423-55-4 | 99.83% | 169.13 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY42145500MG
Encompass_Preferred
3-Maleimidopropionic acid is an alkyl chain-based PROTAC linker used in the synthesis of PROTACs. These compounds facilitate the selective degradation of target proteins by exploiting the intracellular ubiquitin-proteasome system. For research use only.
  • Alkyl chain-based PROTAC linker
  • Facilitates synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-004-04037