Learn More
Medchemexpress LLC 3-Mercaptopropionic acid NHS ester | 117235-10-6 | ≥97.0% | 203.22 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY136159100MG
Encompass_Preferred
3-Mercaptopropanyl-N-hydroxysuccinimide ester is an alkyl/ether-based PROTAC linker. It is used in the synthesis of PROTACs, which are molecules designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. PROTACs achieve this by connecting two different ligands: one for an E3 ubiquitin ligase and the other for the target protein.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Designed to exploit the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-48738