Learn More
Medchemexpress LLC 3-methoxy-5-(pyridin-2-ylethynyl)pyridine | 524924-76-3 | 98.2% | 210.23 | C13H10N2O | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY1251010MM 1ML
Encompass_Preferred
Methoxy-PEPy (3-methoxy-5-(pyridin-2-ylethynyl)pyridine) is a research-use small molecule that acts as a negative allosteric modulator/antagonist of the metabotropic glutamate receptor 5 (mGlu5). It is supplied in solid form and as a 10 mM solution in DMSO (1 mL), and is provided with characterization data including molecular formula C13H10N2O, molecular weight 210.23, and typical purity ~98.2%.
  • Ready-to-use 10 mM solution in DMSO for in vitro assays.
  • Also available as solid in multiple milligram quantities for custom preparations.
  • High purity supports reproducible pharmacology studies.
  • Characterized molecular formula and molecular weight for analytical reference.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-37544