Learn More
Medchemexpress LLC 3-methoxy-N-(1-methylpiperidin-1-ium-4-yl)-4-[[4-(3-oxidanylidene-1,2-dihydroinden-4-yl)oxy]-5-(trifluoromethyl)... | 1227948-02-8 | 98.6% | 555.55 g/mol | C28H29F3N5O4 | 50MG
SDP
Supplier:  Medchemexpress LLC HY12462550MG
Encompass_Preferred
BI-4464 is a potent, highly selective ATP-competitive inhibitor of PTK2 (focal adhesion kinase, FAK) used in biochemical and cell-based studies and as a ligand for proteolysis-targeting chimera (PROTAC) development. It is characterized by a molecular weight of 555.55 g/mol, formula C28H29F3N5O4, and high reported purity suitable for research applications.
Catalog No. 50-002-39378