Learn More
eMolecules​ 26905-40-8 | 3-Methoxybenzylmagnesium chloride, 0.25 M in 2-MeTHF | Synthonix - Stock | MFCD01319896 | 180.910 | C8H9ClMgO | 0.000 | [Cl-].COc1cccc(C[Mg+])c1 | 50ml | 507877493
Supplier:  eMolecules​ 507877493
Encompass_Preferred
3-Methoxybenzylmagnesium chloride, 0.25 M in 2-MeTHF | Synthonix - Stock | 26905-40-8 | MFCD01319896 | 180.910 | C8H9ClMgO | 0.000 | [Cl-].COc1cccc(C[Mg+])c1 | 50ml | 507877493
Catalog No. 50-269-7227