Learn More
Medchemexpress LLC 3-methoxyisoquinoline-1-carboxylic acid | 374917-64-3 | MFCD01845410 | 99.6% | 203.20 | C11H9NO3 | 1 G

Supplier: Medchemexpress LLC HYW0656701G
3-Methoxyisoquinoline-1-carboxylic acid is an organic chemical intermediate used in medicinal chemistry for the synthesis of active compounds. It is supplied for research use only and is not intended for human therapeutic or diagnostic use.
- Use as a drug intermediate for synthesis
- Molecular formula: C11H9NO3
- Molecular weight: 203.20 g/mol
- Purity: 99.6% (HPLC)
- Storage: store at room temperature; in solvent, store at -80°C for long-term preservation
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.