Learn More
Medchemexpress LLC 3-methoxyisoquinoline-1-carboxylic acid | 374917-64-3 | MFCD01845410 | 99.6% | 203.20 g/mol | C11H9NO3 | 100 MG

Supplier: Medchemexpress LLC HYW065670100MG
3-methoxyisoquinoline-1-carboxylic acid is a small-molecule chemical intermediate used for the synthesis of various active compounds and research reagents. It is supplied for research use only and is not intended for human or clinical use.
- Chemical identity: 3-methoxyisoquinoline-1-carboxylic acid
- Cas number: 374917-64-3
- Chemical formula: C11H9NO3
- Molecular weight: 203.20 g/mol
- Typical purity: 99.6% (HPLC, batch COA)
- Storage recommendations: room temperature for solid (recommended up to 3 years); in solvent store at -20°C to -80°C for extended stability
- Intended use: chemical/drug intermediate for research and synthesis
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.