Learn More
Medchemexpress LLC 3-methoxyphenylacetic acid | 1798-09-0 | MFCD00004334 | 99.4% | 166.17 g·mol⁻1 | C9H10O3 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW015343100G
Encompass_Preferred
3-Methoxyphenylacetic acid (CAS 1798-09-0) is an organic acid used as an analytical reagent, reference standard, and synthetic intermediate. It is supplied as a solid and as a DMSO solution in multiple pack sizes and is offered with high reported purity for research and analytical use.
  • Chemical formula: C9H10O3; molecular weight: 166.17 g·mol⁻1.
  • High purity (approximately 99.4%).
  • Available as solid and as 10 mM solution in DMSO.
  • Suitable for use as a reference standard and synthetic intermediate.
  • Pack sizes available from milliliter solutions to multi-hundred gram solids.
Catalog No. 50-003-03675