Learn More
Medchemexpress LLC 3β-methoxypregnenolone | 511-26-2 | 99.7% | 330.50 g·mol-1 | C22H34O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1071165MG
Encompass_Preferred
3β-Methoxypregnenolone is a synthetic derivative of pregnenolone that binds microtubule-associated protein 2 (MAP2), stimulates tubulin polymerization, enhances neurite extension, and demonstrates neuroprotective effects in preclinical studies. Supplied as a high-purity solid for laboratory research use only.
  • Binds microtubule-associated protein 2 and modulates microtubule dynamics.
  • Stimulates tubulin polymerization in vitro.
  • Enhances neurite extension in neuronal models.
  • Shows neuroprotective activity against neurotoxic agents.
  • High purity (~99.7%) suitable for analytical and biological assays.
  • Supplied as 5 mg solid units for laboratory use.
Catalog No. 50-002-39953