Learn More
Medchemexpress LLC 3-methoxytyramine-d4 hydrochloride | 1216788-76-9 | 99.0% | 207.69 | C9H10D4ClNO2 | 10 MG

Supplier: Medchemexpress LLC HY103638S10MG
Deuterated analogue of 3-methoxytyramine supplied as the hydrochloride salt for use as an internal standard, tracer, and analytical reference in quantitative assays (LC-MS, GC-MS, NMR). Supplied in small reference sizes with high purity suitable for method development and calibration.
- Deuterium-labeled hydrochloride salt (d4)
- Purity 99.0%
- Molecular formula C9H10D4ClNO2
- Molecular weight 207.69
- Intended as an internal standard for LC-MS, GC-MS, and NMR
- Available in small reference sizes, including 10 mg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.