Learn More
Medchemexpress LLC 3-methoxytyramine-d4 hydrochloride | 1216788-76-9 | MFCD06658564 | 99.0% | 207.69 g/mol | C9H10D4ClNO2 | 5 MG

Supplier: Medchemexpress LLC HY103638S5MG
3-Methoxytyramine-d4 hydrochloride is the deuterium-labeled analogue of 3-methoxytyramine supplied as a high-purity solid for research and analytical use. It is intended for laboratory applications as a tracer or internal standard in quantitative analytical workflows such as LC-MS, GC-MS, and NMR.
- Purity: 99.0%.
- Molecular weight: 207.69 g/mol.
- Appearance: white to off-white solid.
- CAS number: 1216788-76-9.
- Deuterated (d4) label for use as an internal standard or tracer.
- Available in small pack sizes for analytical work, including a 5 MG pack.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.