Learn More
Medchemexpress LLC 3-methoxytyramine hydrochloride | 1477-68-5 | MFCD00012902 | >95.0% | 203.67 | C9H14ClNO2 | 500 MG

Supplier: Medchemexpress LLC HY103638500MG
3-Methoxytyramine hydrochloride is a dopamine metabolite used in research that can activate trace amine-associated receptor 1 (TAAR1). Provided as a research-grade analytical standard, it is intended for laboratory applications and should be stored sealed, away from moisture and light, under low-temperature conditions to preserve stability.
- acts as a biochemical standard and dopamine metabolite for research applications.
- reported to activate trace amine-associated receptor 1 (TAAR1) for pharmacological studies.
- molecular formula C9H14ClNO2 and molecular weight 203.67 support analytical identification.
- recommended storage sealed and refrigerated or frozen to maintain stability.
- solubility data provided for preparing stock solutions at common concentrations.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.