Learn More
Medchemexpress LLC 3-methyl-2-[(3S)-3-methylpiperidine-1-carbonyl]-4-propan-2-yl-1,2-oxazol-5-one | 654059-21-9 | 99.8% | 266.34 g·mol⁻¹ | C14H22N2O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10205650MG
Encompass_Preferred
A potent, selective small-molecule inhibitor of hormone-sensitive lipase (HSL) intended for biochemical and cellular research. The compound shows an IC50 of 0.023 μM and is supplied as a high-purity solid suitable for assays that probe lipolysis and metabolic pathways. Follow recommended cold-storage conditions to maintain stability and activity.
  • Potent HSL inhibition (IC50 0.023 μM), enabling low-concentration studies.
  • High purity (~99.8%), reducing assay impurities.
  • Suitable for biochemical and cellular assays probing lipolysis pathways.
  • Stable as a powder under recommended cold storage for extended shelf life.
  • Soluble in common organic solvents such as DMSO for formulation flexibility.
Catalog No. 50-002-48514