Learn More
Medchemexpress LLC 3-methyl-2-quinoxalinone | 14003-34-0 | 99.6% | 160.17 | C9H8N2O | 25 G

Supplier: Medchemexpress LLC HY11843625G
3-Methyl-2-quinoxalinone is a heterocyclic synthetic intermediate used in pharmaceutical and organic synthesis. It is supplied as a white crystalline powder with chemical formula C9H8N2O and a molecular weight of 160.17 g/mol. The material is provided in laboratory-scale packaging and is intended for research use.
- High purity: 99.62%.
- Chemical formula C9H8N2O.
- Molecular weight 160.17 g/mol.
- Suitable as a synthetic intermediate for pharmaceutical synthesis.
- Supplied in laboratory quantities, including 25 G packaging.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.