Learn More
Medchemexpress LLC 3-methyl-4-nitrophenol | 2581-34-2 | 99.7% | 153.14 g/mol | C7H7NO3 | 50 G

Supplier: Medchemexpress LLC HYW04233750G
3-methyl-4-nitrophenol is an organic nitrophenol used as an analytical reagent and research chemical, commonly employed in environmental analysis and method development.
- High purity suitable for analytical use.
- Molecular weight 153.14 g/mol and formula C7H7NO3.
- Available in multiple pack sizes including 50 g.
- Supplied as a solid reagent for laboratory applications.
- Commonly used as an analytical standard in environmental studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.