Learn More
Medchemexpress LLC 3-methyl-4-nitrophenol | 2581-34-2 | 99.7% | 153.14 g/mol | C7H7NO3 | 500 G

Supplier: Medchemexpress LLC HYW042337500G
3-methyl-4-nitrophenol is a research chemical and environmental contaminant used in toxicology and environmental degradation studies. It is a degradation product of the insecticide fenitrothion and has been reported to induce apoptosis in nasal epithelial cells and to be toxic to the female reproductive system.
- Molecular formula: C7H7NO3
- Cas number: 2581-34-2
- Purity: 99.7%
- Molecular weight: 153.14 g/mol
- Appearance: yellow to brown solid
- Solubility: DMSO 100 mg/mL (requires ultrasonic agitation)
- Package quantity: 500 g
- Storage: store solutions at -20°C to -80°C for long-term stability
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.