Learn More
Medchemexpress LLC 3-methyl-4-nitrophenol | 2581-34-2 | MFCD00007333 | 99.7% | 153.14 | C7H7NO3 | 1 KG

Supplier: Medchemexpress LLC HYW0423371KG
3-methyl-4-nitrophenol (CAS 2581-34-2) is a research-grade small molecule used as a reference compound and reagent in analytical and biochemical assays. It is supplied as a high-purity solid suitable for laboratory use.
- Purity: 99.7%
- Molecular weight: 153.14 g/mol
- Chemical formula: C7H7NO3
- Available pack sizes including 1 kg
- Intended for research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.