Learn More
Medchemexpress LLC 3-methyl-5-nitropyridine | 6960-20-9 | MFCD04114214 | 100.0% | 138.13 g/mol | C6H6N2O2 | 10 G

Supplier: Medchemexpress LLC HYW01579610G
3-Methyl-5-nitropyridine (CAS 6960-20-9) is an organic intermediate used in the synthesis of pharmaceutical and specialty chemical compounds. Supplied as a powdered reagent, it is intended for research and synthetic applications.
- Chemical intermediate for synthesis of active compounds.
- Molecular formula C6H6N2O2; molecular weight 138.13 g/mol.
- High reported purity (approximately 99.96%).
- Available in multiple pack sizes, including 10 g.
- Powder; store at -20°C (up to 3 years) or 4°C (up to 2 years) as recommended.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.