Learn More
Medchemexpress LLC 3-methyl-D-histidine-d3 hydrochloride | 208.66 | C7H9D3ClN3O2 | 10 MG

Supplier: Medchemexpress LLC HYW017007SB10MG
3-Methyl-D-histidine-d3 hydrochloride is a deuterium-labeled hydrochloride salt of 3-methyl-D-histidine supplied as a powdered reference standard for metabolic and proteomic research. It is commonly used as an internal standard in mass spectrometry and quantitative assays to measure protein-derived 3-methylhistidine and investigate muscle protein turnover.
- Deuterium-labeled internal standard for mass spectrometry.
- Hydrochloride salt supplied as a stable powder.
- High solubility in DMSO (≈100 mg/mL) for convenient stock preparation.
- Molecular weight approximately 208.66 g/mol.
- Store at 4°C, protect from moisture and light.
- Available in small-quantity vials suitable for standardization and method development.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.