Learn More
Medchemexpress LLC 3-methyl-D-histidine-d3 hydrochloride | 208.66 g/mol | C7H12ClN3O2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW017007SB1MG
Encompass_Preferred
3-methyl-D-histidine-d3 hydrochloride is a deuterium-labeled analog of 3-methyl-D-histidine provided as the hydrochloride salt. It is used as an isotopically labeled internal standard or analytical reference in quantitative LC-MS/MS and metabolic research, representing the endogenous residue found in actin and myosin.
  • Isotope-labeled (d3) for accurate mass spectrometry quantitation.
  • Hydrochloride salt for improved solubility and stability.
  • Suitable as an internal standard for LC-MS/MS and metabolic assays.
  • Supplied in small reference quantities for analytical use.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-003-00614