Learn More
Medchemexpress LLC 3-methyl-L-histidine-d3 hydrochloride | MFCD00083658 | 99.7% | 208.66 | C7H9D3ClN3O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW017007SA10MG
Encompass_Preferred
3-Methyl-L-histidine-d3 hydrochloride is a deuterium-labeled internal standard designed for accurate quantification of 3-methylhistidine in biological and food samples by LC-MS/MS. Supplied as the hydrochloride salt with high isotopic enrichment, it provides consistent recovery and reliable peak identification for metabolomics and proteolysis studies. Molecular weight 208.66; formula C7H9D3ClN3O2.
  • Stable isotope-labeled internal standard (d3) for mass spectrometry.
  • Hydrochloride salt form for improved stability and handling.
  • High isotopic enrichment and chemical purity for accurate quantitation.
  • Suitable for metabolomics, pharmacokinetics, and proteolysis assays.
  • Offered in small pack sizes for analytical workflows.
Catalog No. 50-003-00652