Learn More
Cayman Chemical 3-MethyladenIn 100mg
SDP
Supplier:  Cayman Chemical 13242100 MG
Encompass_Preferred
A specific inhibitor of PI3K activity and a widely used inhibitor of the autophagy; at 5 mM, inhibits protein degradation in rat hepatocytes by 65% and blocks class I, II, and III PI3Ks, including some downstream targets
Catalog No. 50-295-2265