Learn More
Medchemexpress LLC 3-Methyladenine-d3 | 110953-39-4 | 98.98% | 152.17 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY19312S1MG
Encompass_Preferred
3-Methyladenine-d3 is the deuterium labeled form of 3-Methyladenine, a PI3K inhibitor. This compound is a commonly used inhibitor of autophagy due to its inhibitory effect on class III PI3K. It can be used as a tracer and as an internal standard for quantitative analysis by various methods. Deuterium substitution has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer for quantitation during drug development
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Inhibits class III PI3K, making it an inhibitor of autophagy
  • Stable heavy isotopes can affect pharmacokinetic and metabolic profiles
Catalog No. 50-003-85566