Learn More
Medchemexpress LLC 3-Methyladenine-d3 | 110953-39-4 | 99.0% | 152.17 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY19312S5MG
Encompass_Preferred
3-Methyladenine-d3 is a deuterium-labeled 3-Methyladenine. 3-Methyladenine (3-MA) acts as a PI3K inhibitor and is widely used as an autophagy inhibitor due to its inhibitory effect on class III PI3K.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-85807