Learn More
Medchemexpress LLC 3-Methylazetidin-3-ol hydrochloride | 124668-46-8 | MFCD11100994 | ≥97.0% | C4H10ClNO | 50 G

Supplier: Medchemexpress LLC HY5933350G
3-Methylazetidin-3-ol hydrochloride is a drug intermediate used for the synthesis of various active compounds. It is intended for research use only and not for patient use. This compound has a molecular weight of 123.58 and a chemical formula of C4H10ClNO, appearing as a white to off-white solid.
- Drug intermediate for synthesis of active compounds
- For research use only
- Solid appearance
- White to off-white color
- Purity of ≥97.0%
- Store at room temperature for 3 years
- In solvent, store at -80°C for 2 years or -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.