Learn More
Medchemexpress LLC 3-Methylbenzoic acid | 99-04-7 | 99.9% | C8H8O2 | 1 KG
SDP
Supplier:  Medchemexpress LLC HY785711KG
Encompass_Preferred
3-Methylbenzoic acid is a weakly acidic compound and serves as an important intermediate in the synthesis of various chemicals, drugs, and agrochemicals, including the insect repellent DEET.
  • Weakly acidic compound
  • Important intermediate for chemical, drug, and agrochemical synthesis
  • Used in insect repellent DEET production
  • Appearance is solid
  • Color is white to off-white
Catalog No. 50-004-33236