Learn More
Medchemexpress LLC 3-Methylbenzoic acid | 99-04-7 | 99.9% | C8H8O2 | 500 G

Supplier: Medchemexpress LLC HY78571500G
3-Methylbenzoic acid is a weakly acidic compound that serves as an important intermediate for the synthesis of various chemicals, drugs, and agrochemicals, including the insect repellent DEET.
- Weakly acidic compound
- Important intermediate for chemical synthesis
- Used in agrochemicals and drugs
- For research use only
- Purity of 99.9%
- White to off-white solid appearance
- Store powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.