Learn More
Medchemexpress LLC HY-113410 500mg Medchemexpress, 3-Methylglutaric acid CAS:626-51-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY113410500MG
Encompass_Preferred
Medchemexpress, HY-113410 500mg 3-Methylglutaric acid CAS:626-51-7 3-Methylglutaric acid is a leucine metabolite. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9252