Learn More
Medchemexpress LLC 3-Methylglutaric acid | 626-51-7 | 98.0% | 146.14 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11341010MM/1ML
Encompass_Preferred
3-Methylglutaric acid is a non-selective inhibitor of mitochondrial function and Na+, K+-ATPase. It induces reactive oxygen species (ROS) generation, leading to oxidative damage and inhibiting mitochondrial redox potential and ion pump function of cell membranes. This product can be used for studying neuropathological mechanisms of metabolic diseases and the role of oxidative stress-mediated neuronal damage in neurodegeneration.
  • Non-selective inhibitor of mitochondrial function and Na+, K+-ATPase.
  • Induces reactive oxygen species (ROS) generation.
  • Can cause oxidative damage.
  • Inhibits mitochondrial redox potential and ion pump function of cell membranes.
  • Appears as a white to off-white solid.
  • Useful for studying neuropathological mechanisms of metabolic diseases.
  • Aids in understanding the role of oxidative stress-mediated neuronal damage in neurodegeneration.
Catalog No. 50-004-36799