Learn More
Medchemexpress LLC (3-Methyloxiran-2-yl)methanol | 872-38-8 | MFCD00033455 | 95.0% | 88.11 g/mol | C4H8O2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW310318500MG
Encompass_Preferred
(3-Methyloxiran-2-yl)methanol is an epoxide-containing small molecule used primarily as a drug intermediate and reference compound in medicinal chemistry and organic synthesis. It is a low-molecular-weight, polar epoxide alcohol employed to introduce a 2,3-epoxybutanol fragment into target molecules and for ring-opening transformations.
  • Epoxide alcohol functional group for ring-opening chemistry.
  • Useful building block for medicinal chemistry and synthesis of active compounds.
  • Reported purity approximately 95% (supplier specification).
  • Molecular weight 88.11 g/mol and formula C4H8O2.
  • Available in small research-scale pack sizes suitable for lab use.
  • Provided with SDS, datasheet, and COA documentation for handling and verification.
Catalog No. 50-002-82517