Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Methylpyrazole | 1453-58-3 | 99.5% | C4H6N2 | 5 G
SDP

Supplier:  Medchemexpress LLC HY660545G

Encompass_Preferred

This product is 3-Methylpyrazole, a chemical compound primarily used as a nitrification inhibitor in soil. It also demonstrates the ability to inhibit the metabolism of methylazoxymethanol (MAM) by rat liver microsomes.

  • Used as a nitrification inhibitor in soil
  • Inhibits MAM metabolism by rat liver microsomes
  • Appearance: Light yellow to reddish brown
  • Purity (HPLC): 99.5%
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in H2O (25 mg/mL) and PBS (50 mg/mL)

Catalog No. 50-004-33851


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.