Learn More
Medchemexpress LLC 3-Methyluric Acid-13C4,15N3 | 1173019-10-7 | 99.5% | C213C4H6N15N3O3 | 2 MG
SDP
Supplier:  Medchemexpress LLC HY141917S2MG
Encompass_Preferred
3-Methyluric Acid-13C4,15N3 is an isotope-labeled compound, specifically a 13C and 15N labeled form of 3-Methyluric Acid. Stable heavy isotopes of elements like carbon and nitrogen are incorporated into drug molecules, primarily serving as tracers for quantitation during the drug development process.
  • Labeled with 13C and 15N isotopes
  • Suitable as a tracer for quantitation in drug development
  • Contributes to understanding pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-53293