Learn More
Medchemexpress LLC 3-Methylxanthine (standard) | 1076-22-8 | 99.58% | C6H6N4O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY50723R100MG
Encompass_Preferred
3-Methylxanthine (Standard) is the analytical standard of 3-Methylxanthine, intended for research and analytical applications. It is a xanthine derivative and a cyclic guanosine monophosphate (GMP) inhibitor, with an IC50 of 920 μM on guinea-pig isolated trachealis muscle. This analytical standard is used as a reference standard supplied for assay.
  • Commonly utilized in qualitative research experiments
  • Commonly utilized in quantitative research experiments
  • Commonly utilized in methodological research experiments
  • Used in HPLC
  • Used in GC
  • Used in MS
Catalog No. 50-003-59127