Learn More
Selleck Chemical LLC 3-n-Butylphathlide S9263-1MG
SDP
Supplier:  Selleck Chemical LLC S92631MG
Encompass_Preferred
3-n-Butylphathlide initially isolated from the seeds of Apium graveolens Linn has shown significant neuroprotective effects
Catalog No. 50-000-23787