Learn More
Medchemexpress LLC 3-(naphthalene-1-carbonylamino)pyrazine-2-carboxylic acid | 2755712-12-8 | 99.5% | 293.28 | C16H11N3O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15063710MG
Encompass_Preferred
Mab Aspartate Decarboxylase-IN-1 is a small-molecule research reagent used to inhibit aspartate decarboxylase (PanD) for biochemical and microbiological studies, including investigation of β-alanine biosynthesis and antibacterial effects against Mycobacterium abscessus.
  • Potent PanD inhibitor (IC50 56.3 μM).
  • Demonstrates antibacterial activity against Mycobacterium abscessus (IC50/MIC in the mM range).
  • High chemical purity (99.5%).
  • Solid, white to off-white appearance suitable for standard laboratory handling.
  • Stable when stored sealed at 4°C; in solvent, -80°C for 6 months or -20°C for 1 month.
  • Well-characterized chemical data: formula C16H11N3O3; molecular weight 293.28.
Catalog No. 50-002-86419