Learn More
Sigma Aldrich Fine Chemicals Biosciences 3-Nitrobenzyl alcohol suit5G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 731485G
Encompass
3-Nitrobenzyl alcohol (3-NBA) is most commonly used matrix for spectrometry methods. It helps in the analysis of porphyrins cyclic peptides alkali metal cationized fatty acids for collisional activation and also certain organoruthenium compounds.
Catalog No. 50-289-7164