Learn More
Medchemexpress LLC 3-Nitropropanoic acid | 504-88-1 | 99.9% | 119.08 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01287510MM/1ML
Encompass_Preferred
3-Nitropropanoic acid is an irreversible and orally active inhibitor of succinate dehydrogenase. It demonstrates potent antimycobacterial activity with a MIC value of 3.3 μM and can induce cell apoptosis. This compound is suitable for various research applications, including cellular studies and induced disease models.
  • Irreversible and orally active succinate dehydrogenase inhibitor
  • Potent antimycobacterial activity
  • Induces cell apoptosis and autophagy
  • Disrupts mitochondrial morphology in SH-SY5Y cells
  • Used in induced disease models, such as Huntington's Disease
  • Soluble in DMSO and water
Catalog No. 50-004-54807