Learn More
Medchemexpress LLC 3-Nitropropanoic acid | 504-88-1 | 99.9% | 119.08 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW012875500MG
Encompass_Preferred
3-Nitropropanoic acid is an irreversible and orally active inhibitor of succinate dehydrogenase. It exhibits potent antimycobacterial activity with a MIC value of 3.3 μM and can induce cell apoptosis.
  • Irreversible and orally active inhibitor of succinate dehydrogenase.
  • Potent antimycobacterial activity.
  • Can induce cell apoptosis.
  • Induces autophagy and disrupts mitochondrial morphology in SH-SY5Y cells.
  • Induces oxidative stress and apoptosis of granulosa cells in geese.
  • Induces cell death in cultured rat hippocampal neurons.
  • Induces oxidative stress and increases lipid peroxidation in brain regions of the Wistar rat.
  • Evokes seizures in mice.
  • Induction of Huntington's Disease (HD) model in rats.
Catalog No. 50-004-54893