Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Nitropyrazole | 26621-44-3 | 113.08 | 100 G
SDP

Supplier:  Medchemexpress LLC HYW002457100G

Encompass_Preferred

3-Nitropyrazole is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also a nitrogen-containing heterocyclic compound, which can be used in the synthesis of other active compounds. The nitro group of 3-Nitropyrazole readily undergoes nucleophilic substitution reactions.

  • Used as a biological material or organic compound
  • Nitrogen-containing heterocyclic compound
  • Can be used in the synthesis of other active compounds
  • Nitro group readily undergoes nucleophilic substitution reactions
  • Appearance: White to light yellow solid
  • Purity: 99.98%
  • Molecular weight: 113.08
  • Storage: Powder at -20°C for 3 years; 4°C for 2 years; in solvent at -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-45887


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.