Learn More
Medchemexpress LLC 3-Nitropyrazole | 26621-44-3 | 113.08 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00245750G
Encompass_Preferred
3-Nitropyrazole is a biochemical reagent suitable for life science-related research as a biological material or organic compound. It is also a nitrogen-containing heterocyclic compound that can be utilized in the synthesis of other active compounds. This product is for research use only and is not sold to patients.
  • The nitro group readily undergoes nucleophilic substitution reactions
Catalog No. 50-004-45792