Learn More
Abcam 3-Nitrotyrosine ELISA Kit - Intracellular, 10 x 96 Tests
Supplier:  Abcam AB31690510X96TESTS
Encompass_Preferred
3-Nitrotyrosine ELISA Kit - Intracellular is a single-wash 90-min Simplestep used to quantify 3-Nitrotyrosine with a sensitivity of 40.3 ng/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Competitive ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-271-1634
Add to Cart
Request a Quote