Learn More
Medchemexpress LLC 3'-O-(2-Methoxyethyl)adenosine | 303197-30-0 | C13H19N5O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1524985MG
Encompass_Preferred
3'-O-(2-Methoxyethyl)adenosine is an adenosine analogue that functions as a smooth muscle vasodilator and has shown to inhibit cancer progression. It is intended for research use only.
  • Adenosine analogue
  • Functions as smooth muscle vasodilator
  • May impede cancer progression
  • Purity: 99.87%
Catalog No. 50-003-72366