Learn More
Medchemexpress LLC 3'-O-(2-Methoxyethyl)guanosine | 256224-03-0 | C13H19N5O6 | 1 ML

Supplier: Medchemexpress LLC HY15249310MM/1ML
3'-O-(2-Methoxyethyl)guanosine is a guanosine analogue exhibiting immunostimulatory activity. It induces type I interferons, leading to antiviral effects. Its functional activity is dependent on the activation of Toll-like receptor 7 (TLR7).
- Immunostimulatory activity
- Induces type I interferons for antiviral effects
- Activates Toll-like receptor 7 (TLR7)
- Molecular weight: 341.32
- Purity: 98.90%
- Appearance: Solid
- Color: White to off-white
- Stable for long-term storage as powder at -20°C (3 years) or 4°C (2 years)
- Stable for short-term storage in solvent at -80°C (6 months) or -20°C (1 month)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.