Learn More
Apexbio Technology LLC 3'-O-(2-nitrobenzyl)-2'-dATP 10ul (100 mM)
SDP
Supplier:  Apexbio Technology LLC B81230.01
Encompass_Preferred
3p-O-2-nitrobenzyl-2p-dATP is a photosensitive nucleotide analog It is chemically modified with a 2-nitrobenzyl group at the 3 -hydroxyl position enabling controlled activation upon exposure to irradiation at specific wavelengths 3p-O-2-nitrobenzyl-2p-dATP exerts its biological activity primarily through photolabile removal of the 2-nitrobenzyl protecting group which generates a free 3 -OH group and allows for enzymatic DNA polymerization Based on these properties 3p-O-2-nitrobenzyl-2p-dATP holds research potential in sequence-controlled DNA synthesis construction of high-resolution nucleic acid microarrays nucleic acid synthetic technologies and studies of spatiotemporally controlled gene expression and polymerase-mediated nucleotide incorporation
Catalog No. 50-000-88023