Learn More
Apexbio Technology LLC ddUTP 50ul (100 mM)
SDP
Supplier:  Apexbio Technology LLC B81380.05
Encompass_Preferred
ddUTP is a synthetic nucleotide analog designed to terminate DNA chain elongation by DNA polymerases ddUTP exerts its biological activity primarily through incorporation into nascent DNA strands where the absence of a 3 -hydroxyl group prevents further phosphodiester bond formation and subsequent nucleotide addition Based on these pharmacological properties ddUTP holds research potential in DNA sequencing methodologies including Sanger sequencing and in termination-based DNA labeling experiments for mapping genomic sequences and structural features
Catalog No. 50-000-91167