Learn More
Medchemexpress LLC 3-O-Acetylhamaudol | 30358-88-4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN68921MG
Encompass_Preferred
3-O-Acetylhamaudol is a chemical compound with CAS Number 30358-88-4. This product should be stored properly to maintain its stability and efficacy.
  • Keep container tightly sealed in a cool, well-ventilated area.
  • Keep away from direct sunlight and sources of ignition.
  • Recommended storage temperature: 4°C, sealed storage, away from moisture and light.
  • If in solvent, store at -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture and light).
  • Shipping at room temperature is acceptable if less than 2 weeks.
Catalog No. 50-004-17903